Limb Lock, Mortech
Supplier: Mortech Manufacturing
BC012
76549-596EA 559.44 USD
76549-596
Limb Lock, Mortech
Ergonomic Supports Ankle Supports
Exclusively offered by Mopec, this simple self-adjusting design provides a safe and steady method to secure all limb specimens during any autopsy or pathology dissection.
  • Heavy gauge stainless steel
The Limb Lock’s unique notched V-shape offers all the benefits of a second set of hands. Multiple units can be utilized simultaneously for larger amputation specimens, and the unit can be easily cleaned with all standard disinfectants.