Solar Powered Hanging Scale, Mortech
Supplier: Mortech Manufacturing
BB067
76549-590EA 971.01 USD
76549-590
Solar Powered Hanging Scale, Mortech
Balances and Scales
The model BB067 scale is easy to use, gives instantaneous weight readouts, and is completely solar powered so you'll never have to worry about battery replacement.
  • Never replace a battery – completely solar powered
  • Has the ability to zero tare bow, ring and pan
  • Digital speed for instantaneous weight readouts
  • Pinpoint accuracy to 0.02 lb / 0.01 kg
  • 30 lb/15 kg capacity for versatile weighing use
  • Easy to operate – no buttons to press
Exclusive solar powered design for optimal efficiency.