Drain Bucket, Plastic, Mortech
Supplier: Mortech Manufacturing
BA102
76549-552EA 70.84 USD
76549-552
Drain Bucket, Plastic, Mortech
Buckets
This quality utility bucket and kickstand can be used for a variety of facility needs.
  • Graduated in pints, quarts, gallons, and liters
  • Meets FDA standards
  • 14 quart total capacity
This injection-molded plastic bucket is designed to resist most acids, alkalies, paint, chemicals, and cleaning compounds.