Griffin Style Beakers, Mortech
Supplier: Mortech Manufacturing
BA081 BA082 BA083 BA084
76549-536EA 107.85 USD
76549-536 76549-538 76549-540 76549-542
Griffin Style Beakers, Mortech
Beakers
Beakers with smooth finish for easy cleaning
  • Made of all stainless steel, seamless construction
  • With pouring spout
  • Available in multiple sizes