Headrest, Solid Aluminum, Mortech
Supplier: Mortech Manufacturing
BA030
76549-514EA 1058.86 USD
76549-514
Headrest, Solid Aluminum, Mortech
Ergonomic Supports Shoulder Supports
Secures cranium during all autopsy and/or mortuary procedures.
  • Adjustable up to six positions
  • Easily cleaned with soap and water
  • Wide base legs allows for more table contact for added stability and less slippage