Scalpel Handle, Stainless Steel No. 4L, Premium Grade, Mortech
Supplier: Mortech Manufacturing
AJ201
76549-488EA 209.55 USD
76549-488
Scalpel Handle, Stainless Steel No. 4L, Premium Grade, Mortech
Scalpels and Scalpel Assemblies Scalpel Handles
High-quality #4 long scalpel handle, manufactured in Germany, can be used with #20 thru #23 blades. Stainless steel.
  • Premium grade stainless steel