Scalpel Handle, Stainless Steel, No. 3, Mortech
Supplier: Mopec
AJ140
76549-446EA 40 USD
76549-446
Scalpel Handle, Stainless Steel, No. 3, Mortech
Scalpels and Scalpel Assemblies Scalpel Handles
#3 ridged surface, stainless steel scalpel handle for #10 thru #15 blades. This tool is durable and reusable.
  • Stainless steel
  • Fits #10, #11, #12, #13, #14 and #15 blades