Scalpel Handle, Molded Grip, No. 6, Mortech
Supplier: Mortech Manufacturing
AJ126
76549-430EA 121.7 USD
76549-430
Scalpel Handle, Molded Grip, No. 6, Mortech
Scalpels and Scalpel Assemblies Scalpel Handles
This molded handle has a smooth grip for comfortable post-mortem dissection. Fits blade sizes #20 – #23.
  • Premium grade stainless steel with ergonomic plastic handle
  • Fits #20 – #23 blades
It can easily be cleaned and autoclaved for long-lasting life.