Scalpel Blades, Post Mortem, Round Tip, PM40, Mortech
Supplier: Mortech Manufacturing
AJ124
76549-426EA 120.17 USD
76549-426
Scalpel Blades, Post Mortem, Round Tip, PM40, Mortech
Scalpels and Scalpel Assemblies Scalpel Blades
This post mortem rounded tip blade is ideal for most cutting procedures.
  • Carbon steel
  • Pack of 10
  • Compatible with AJ125 handle