Blade Remover, Disposable, Mortech
Supplier: Mortech Manufacturing
AJ116
76549-414EA 115.52 USD
76549-414
Blade Remover, Disposable, Mortech
Blade Removers and Dispensers
Designed to safely remove dull contaminated blade from scalpel handle.
  • Pack of 10
  • Autoclavable
  • Holds 100 blades each
  • Not compatible with double sided blades
Simply insert and press handle to dispense old blade safely into sealed container.