Chalk Calvaria Elevator, Mortech
Supplier: Mortech Manufacturing
AH200
76549-380EA 380.65 USD
76549-380
Chalk Calvaria Elevator, Mortech
Retractors
The Chalk Calvaria Elevator is designed with both function and comfort in mind. The penetrating edge is only 10 mm long which prevents over penetration of the cranial cavity and laceration to the brain. The heavy-duty strike plate and handle are mounted at a right angle to the penetrating edge. This makes the use of a mallet easier while holding the tool and allows the user to apply torque to remove the calvaria.
  • Stainless steel
With the Chalk Calvaria Elevator any discomfort from the torque method is eliminated, which in turn will reduce user injury.