Knife Sharpener, Variable Speed, Mortech
Supplier: Mortech Manufacturing
AH032 AH033
76549-332EA 3024.13 USD
76549-332 76549-334
Knife Sharpener, Variable Speed, Mortech
Knives
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge. The honing wheels are mounted on parallel shafts that rotate in opposite directions.
  • Stainless steel case
Available in US and International voltage.
Ordering information: Replacement honing wheels available (See model AH034).