Knife, Gannetts Post Mortem Dissecting, Mortech
Supplier: Mortech Manufacturing
AH015
76549-308EA 53.86 USD
76549-308
Knife, Gannetts Post Mortem Dissecting, Mortech
Knives
The broad non-flexible blade is perfect for cutting dense tissue. The top edge can be struckwith a mallet to hammer through hard tissue and/or bone.
  • Heavy duty stainless steel
  • Extra thick cutting edge