Bone Rongeurs, Cleveland, No. 4A, Mortech
Supplier: Mortech Manufacturing
AG034
76549-238EA 70.84 USD
76549-238
Bone Rongeurs, Cleveland, No. 4A, Mortech
Bone Cutters and Rongeurs
Cleveland rongeurs are designed to cut, sculpt, and trim soft or hard bones. Satin surface to reduce glare.
  • German grade satin stainless steel