Bone Chisel, Brunetti’s, Mortech
Supplier: Mortech Manufacturing
AG015 AG016
76549-214EA 184.9 USD
76549-214 76549-216
Bone Chisel, Brunetti’s, Mortech
Chisels
The Brunetti's chisels are specifically designed to score, cut, scrape, clean and sculpt around the spinal cord.
  • Standard grade stainless steel
Available in right handed and left handed options.