Dissection Tool, Light Duty, Mortech
Supplier: Mortech Manufacturing
AD120
76549-122EA 16.67 USD
76549-122
Dissection Tool, Light Duty, Mortech
Retractors
Ideal for delicate and light duty post-mortem dissection procedures.
  • Dual prong, sharp narrow
Available in a variety of thin to wide spaced curved prongs for gently isolating and retracting fascia, ligament tissue, nerve isolation, nerve teasing and spinal cord dissection. Serrated total perimeter grip assures secure tissue handling.