Stapler, Postmortem, Mortech
Supplier: Mortech Manufacturing
AD046
76549-070EA 95.49 USD
76549-070
Stapler, Postmortem, Mortech
Staplers Staple Guns
Postmortem stapler provides a safe, fast and efficient option for skin closure.
  • Staples 0.25"
  • Pre-loaded with 35 staples
  • Disposable
Reduces needle and sharps exposures. Arrow guide assists in accurate staple placement. It also features an easy-to-read staple counter.