Bullet Probe, 22 Caliber, Mortech
Supplier: Mopec
AD022
76549-044EA 184.9 USD
76549-044
Bullet Probe, 22 Caliber, Mortech
Dissection Needles
Designed for tracking bullet path and angle of entry.
  • Stainless steel
Blunt probing end has a rounded tip to minimize tissue damage.