Forceps, Micro Grooved Tissue, Curved, Mortech
Supplier: Mortech Manufacturing
AB154
76548-860EA 473.13 USD
76548-860
Forceps, Micro Grooved Tissue, Curved, Mortech
Forceps
Unique octagon handle shape provides a broad area for the finger to grasp for secure, yet delicate post-mortem dissections.
  • Premium grade stainless steel
Micro fine tips for precise extraction.