Forceps, Micro Iris Tissue, Mortech
Supplier: Mortech Manufacturing
AB150 AB151
76548-852EA 622.65 USD
76548-852 76548-854
Forceps, Micro Iris Tissue, Mortech
Forceps
Unique octagon handle shape provides a broad area for the finger to grasp for secure, yet delicate post-mortem dissections.
  • Premium grade stainless steel
  • 1×2 teeth tips
Available with straight or curved edges.