Forceps with Imprinted Ruler, Mortech
Supplier: Mortech Manufacturing
AB090 AB091
76548-768EA 126.32 USD
76548-768 76548-770
Forceps with Imprinted Ruler, Mortech
Forceps
Serrated forceps imprinted with metric guides on the tips. This instrument is made for precise and accurate post-mortem dissections.
  • Stainless steel
  • Ruler up to 50 mm on both forcep lengths