Forceps, CutMATE, Mortech
Supplier: Mortech Manufacturing
AB071 AB072 AB073 AB074 AB075
76548-750EA 1448.84 USD
76548-750 76548-752 76548-754 76548-756 76548-758
Forceps, CutMATE, Mortech
Forceps
CutMATE forceps were developed to overcome the variability of tissue specimen thickness. They will allow cutting consistent tissue specimen thickness of 2, 3, or 4 mm.
  • 304 stainless steel
  • Fits all plastic universal cassettes
The unique 'trident-like' design of the 35 mm long ergonomically angled prongs, ensures uniform tissue thickness cutting of tissue lengths that will fit standard cassettes. The inside surface of the prongs is specially textured to provide the extra grip needed for stabilizing and slicing difficult and hard to hold specimens.