Forceps, Ergonomic, Mortech
Supplier: Mortech Manufacturing
AB054 AB056 AB057 AB058
76548-728EA 118.63 USD
76548-728 76548-732 76548-734 76548-736
Forceps, Ergonomic, Mortech
Forceps
Unique perforated paddle handles for secure slip-free grip. Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
  • Stainless steel
  • Broad grip
Available with smooth or serrated tips.