Forceps, Specimen Paddle, Mortech
Supplier: Mortech Manufacturing
AB042 AB043 AB064
76548-706EA 120.17 USD
76548-706 76548-708 76548-746
Forceps, Specimen Paddle, Mortech
Forceps
Paddle forceps hold all types of tissue with the secure grip needed to accomplish thin sections for analysis. Thin sections are essential for proper infiltration of reagents prior to study. Great for training residents and technicians on thin section technique, cutting thin sections of fatty tissue like breast tissue and cutting even thickness sections of any type tissue.
  • Standard grade stainless steel
  • Ribbed grip
  • Paddle ends
Available in a variety of paddle types.