Forceps, Iris Tissue, Full Curved, 1×2 Teeth, Mortech
Supplier: Mortech Manufacturing
AB038
76548-698EA 41.54 USD
76548-698
Forceps, Iris Tissue, Full Curved, 1×2 Teeth, Mortech
Forceps
Sharp 1×2 interlocking teeth for secure grip with minimal tissue compression.
  • Standard grade stainless steel
  • Full curve tips with 1×2 teeth
  • Ribbed grip