Forceps, Iris Tissue, Curved, Mortech
Supplier: Mortech Manufacturing
AB036
76548-694EA 32.29 USD
76548-694
Forceps, Iris Tissue, Curved, Mortech
Forceps
Curved tip forceps allow for a tight grip with minimal tissue compression.
  • Standard grade stainless steel
  • Ribbed grip
  • Full curved with serrated pattern tips