Scissors, Titanium, Metzenbaum, Mortech
Supplier: Mortech Manufacturing
AA703 AA704 AA705 AA706
76548-614EA 209.55 USD
76548-614 76548-616 76548-618 76548-620
Scissors, Titanium, Metzenbaum, Mortech
Scissors
Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the top choice for many technicians. Titanium alloy instruments are keep edges sharp for longer. More stain resistant than stainless steel. Metzenbaum scissors have a longer handle to blade ratio and can cut extremely delicate tissue.
  • Standard grade titanium alloy
  • 25% lighter that stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.