Scissors, Titanium, Mayo, Mortech
Supplier: Mortech Manufacturing
AA701 AA702
76548-610EA 195.68 USD
76548-610 76548-612
Scissors, Titanium, Mayo, Mortech
Scissors
Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium alloy instruments keep edges sharp longer than traditional instruments. More stain resistant than stainless steel.
  • Standard grade titanium alloy
  • 25% lighter that stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.