Scissors, Metzenbaum, Straight, Mortech
Supplier: Mortech Manufacturing
AA023 AA024 AA037 AA038 AA123 AA124 AA137 AA138
76548-352EA 40 USD
76548-352 76548-354 76548-380 76548-382 76548-446 76548-448 76548-468 76548-470
Scissors, Metzenbaum, Straight, Mortech
Scissors
These scissors offer a unique combination of slim, blunt tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
  • Stainless steel
  • Straight cutting edge
  • Narrow blunt points
Available in standard or premium grade.