Scissors, Iris, Angled, Mortech
Supplier: Mortech Manufacturing
AA030 AA130
76548-366EA 43.06 USD
76548-366 76548-460
Scissors, Iris, Angled, Mortech
Scissors
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Angled end cutting edge
  • Sharp points
Available in standard or premium grade.