Scissors, Metzenbaum, Curved, Mortech
Supplier: Mortech Manufacturing
AA025 AA026 AA039 AA040 AA125 AA126 AA139 AA140
76548-356EA 41.54 USD
76548-356 76548-358 76548-384 76548-386 76548-450 76548-452 76548-472 76548-474
Scissors, Metzenbaum, Curved, Mortech
Scissors
These scissors offer a unique combination of slim, blunt, curved tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
  • Stainless steel
  • Curved cutting edge
  • Narrow blunt points
Available in standard or premium grade.