Hematoxylin, Vintage Plus

Supplier: StatLab
Warning

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

SL106
76548-216EA 76.19 USD
76548-216
Hematoxylin, Vintage Plus
Hematoxylin staining solutions

Hematoxylin optimized for mucin staining, great for gastroenterology, delivering an intense hue.


  • Uniquely formulated progressive Hematoxylin to provide the end user with sharp and well delin-eated nuclear chromatin staining
  • Formula has a specific affinity for mucin and is a preferred nuclear stain for GI Laboratories
  • A rapid stain recommended for frozen section staining
  • Can be used for specimens in both Histology and Cytology laboratories


Part of the StatLab Vintage stain line.

Manufactured in USA.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR