Eosin Y, Vintage

Supplier: StatLab
Warning

SL101
76548-208EA 54.34 USD
76548-208
Eosin Y, Vintage
Eosin Y

Proprietary, alcoholic and acidified formulation


  • Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
  • Yields a very familiar cytoplasmic staining result


Part of the StatLab Vintage stain line.

Manufactured in USA.

Formula: C₂₀H₈Br₄O₅
MDL Number: MFCD00036189
CAS Number: 15086-94-9
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR