Fixative StatFix™, Statlab
Supplier: StatLab
SF-1
76548-204EA 33.93 USD
76548-204
Fixative StatFix™, Statlab
Stains Microscopy and staining reagents
Buffered alcoholic formalin fixative.
  • Particularly well suited for fatty tissues, rapid diagnostic applications and routine frozen sections
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA