PolarStat Plus™ Embedding Medium, StatLab
Supplier: StatLab
PSP-12
76548-202CS 223.69 USD
76548-202
PolarStat Plus™ Embedding Medium, StatLab
Stains Microscopy and staining reagents
PolarStat Plus™ frozen section embedding medium is a proprietary formulation of water soluble glycols and resins, providing an optimal specimen matrix for Mohs cryostat sectioning, and bonding the specimen securely to the specimen chuck. This formulation freezes at optimum cutting temperatures of -10°C and below. No residue is left on slides during the staining procedure, thereby eliminating undesirable background staining. 12 squeeze bottles/case. Sold by the case.
Packaging: Manufactured in the USA