Paraffin XLT, ParaPro™
Supplier: StatLab
Synonyms:
Wax Paraffin, Paraffin wax
PPXLT
76548-200CS
147.46
USD
76548-200
Paraffin XLT, ParaPro™
Paraffin
A superior mixture of waxes and copolymer alloys. A medium/hard paraffin for infiltration and embedding.
- Produces ultra-thin compression-free ribbons and can be used to cut 2-micron sections without icing
- Provides excellent support for all types of tissue samples while maintaining exceptional clarity
- Melting Point 55°-57°C
- 0.8 kg bag, 10/cs
Part of the StatLab line of ParaPro paraffin.
Manufactured in the USA
CAS Number:
8002-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...