Paraffin SEM, ParaPro™

Supplier: StatLab

Synonyms: Wax Paraffin, Paraffin wax

PPSEM
76548-198CS 126.18 USD
76548-198
Paraffin SEM, ParaPro™
Paraffin

Standard Embedding Media. A softer paraffin.


  • For infiltration and embedding
  • Formulation consists of a low polymer count
  • Improves infiltration and embedding and leads to more efficient deparaffinization
  • Minimizes wrinkles
  • Melting point 54-56°C
  • 0.8 kg bag, 10/cs


Part of the StatLab line of ParaPro paraffin.

Manufactured in the USA

CAS Number: 8002-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR