Paraffin LMP, ParaPro™

Supplier: StatLab

Synonyms: Wax Paraffin, Paraffin wax

PPLMP
76548-196CS 151.38 USD
76548-196
Paraffin LMP, ParaPro™
Paraffin

A medium/hard paraffin. For infiltration and embedding.


  • Consists of a specialized blend of low-molecular-weight paraffin and polymers to reduce melting point
  • The lower melting point protects tissues from excessive temperature during processing
  • Melting point 50°-54° C
  • 1 kg bag, 8/cs


Part of the StatLab line of ParaPro paraffin.

Manufactured in the USA

CAS Number: 8002-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR