Millennia™ Command Adhesion Slides
Supplier: StatLab
MCOMM/W/90
76548-148BX 41.34 USD
76548-148
Millennia™ Command Adhesion Slides
Slides Adhesive Slides
A universal adhesion slide for H&E, IHC, and special stains.
  • Hybrid slide suitable for IHC, H&E, and special staining
  • Optimized coating for strong adhesion to reduce tissue loss and provide consistency of staining
  • Exhibits minimal background staining during all types of staining
  • Hydrophilic, 90 degree corners
  • Compatible with thermal transfer printers
Part of the StatLab Millennia Adhesion Microscope Slide family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.