OptiCut™ Microtome Blades, StatLab Medical Products
Supplier: Avantor
CUT7223
76548-102PK 137.18 USD
76548-102 76548-104
OptiCut™ Microtome Blades, StatLab Medical Products
Microtomes and Microtome Assemblies Microtome Blades and Handles
Extra-sharp blades for precise cutting at the microtome.
  • Precision manufactured to the highest standards for sharpness, consistency and durability
  • A proprietary honing and polishing process ensures consistent sharpness and exceptional durability
Blades can accommodate high volume laboratories. Manufactured in USA.