Formalin concentrate 10%, neutral buffered

Supplier: StatLab
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

BF128
76548-076EA 40.42 USD
76548-076
Formalin concentrate 10%, neutral buffered
Formalin

Formalin concentrate


  • Concentrated NBF to save space
  • Combine 1 part concentrate with four parts water for a ready-to-use solution
  • 10% NBF Formalin Concentrate


Part of the StatLab line of 10% NBF Concentrate.

Manufactured in the USA

Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR