Xylene Substitute XS-3™, Statlab
Supplier: StatLab
7400-1
76548-058CS 106.93 USD
76548-058
Xylene Substitute XS-3™, Statlab
Stains Microscopy and staining reagents
100% aliphatic hydrocarbon for processing and staining.
  • Higher permissible exposure limit than xylene
  • Virtually no odor
  • Flash point 104°F
  • Exceptionally high purity
Part of the StatLab line of clearing agents.
Manufactured in the USA