Methanol ACS

Supplier: StatLab
Danger

40380-1
76548-042EA 23.12 USD
76548-042
Methanol ACS
Methanol

99% absolute Methanol.


  • For general tissue processing as a substitute for ethanol or as a cytology fixative.


Part of the StatLab line of alcohol products that are ACS grade.

Manufactured in the USA

Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR