2-Propanol

Supplier: StatLab
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

36420-1
76548-040CS 106.97 USD
76548-040
2-Propanol
2-Propanol

99% absolute Isopropyl Alcohol.


  • Ideal for small tissue, biopsy, and microwave processing


Part of the StatLab line of alcohol products that are ACS grade.

Manufactured in the USA

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR