Formalin 20%, neutral buffered

Supplier: StatLab
Danger

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

28620-5
76548-036EA 79.88 USD
76548-036
Formalin 20%, neutral buffered
Formalin

20% Neutral Buffered Formalin is intended as a general purpose fixative


  • For all tissues to be examined by light microscopy
  • Solution is often used for surgical and necropsy specimens, and is an excellent fixative for brain tissue


Part of the StatLab line of prefilled bulk containers

Manufactured in the USA

Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR