Nitrical™ Decalcifier, StatLab
Supplier: StatLab
Safe and gentle standard AFIP formulation. Made from semiconductor grade acid and highly purified reagent grade water. Excellent decalcifier for busy laboratories where time is a critical element. Very mild solution that works rapidly with beautiful results. Can be used for all types of specimens from bone marrow to coritcal bone.
Packaging: Manufactured in the USA