Rapid Response™ Lateral Flow Reader, BTNX
Supplier: BTNX INC.
LFA-1.0
76547-874CS 10823.38 USD
76547-874
Rapid Response™ Lateral Flow Reader, BTNX
Drug Tests
The Rapid Response™ lateral flow reader analyzes results and automatically generates and stores test reports from Rapid Response™ Multi-drug test panels.
  • Integrates directly with PateintVuTM and other LIS and EMR systems for seamless workflow, administration and record-keeping
  • Analyzes results within 10 s
  • Saves test results and generates customized test results
  • QR code recognition identifies the patient, sample and test type
  • Flexible operator access, test and method settings
This product is only compatible with Rapid Response™ Multi-drug test panels.