Virex® Plus One-Step Disinfectant Cleaner & Deodorant
Supplier: DIVERSEY INC.
101102925
76547-806CT 233.19 USD
76547-806
Virex® Plus One-Step Disinfectant Cleaner & Deodorant
Cleaning Agents Disinfectants
A one-step, quaternary-based disinfectant cleaner concentrate providing broad spectrum disinfection at 1:256 dilution. 
  • Kills HIV-1 and Influenza Type A in 1 minute
  • Kills Methicillin Resistant Staphylococcus aureus (MRSA), Vancomycin Resistant Enterococcus faecalis (VRE), Klebsiella pneumoniae Carbapenum-resistant (CRKP), Acinetobacter baumannii, and other pathogens in 3 minutes
  • Kills Norovirus in 5 minutes at a 1:128 dilution (1 oz. per gallon)
  • Cleans and disinfects finished floors without dulling gloss
  • Bactericidal: Kills many pathogenic bacteria
  • Inhibits the growth of mold and mildew and their odors. Fungicidal: Kills Trichophyton mentagrophytes (athlete's foot fungus) in 3 minutes. Non-Food Contact Sanitizer in 15 seconds at 1:256
Use in healthcare and other facilities where cleaning and prevention of cross-contamination are critical.
Bactericidal, virucidal and fungicidal. Kills MRSA and VRE.
Meets bloodborne pathogen standards for decontaminating blood and body fluids.
Blue in color with no added fragrance.