Crew® Restroom Floor & Surface SC Non-Acid Disinfectant Cleaner
Supplier: DIVERSEY INC.
101102189
76547-804CT 232.06 USD
76547-804
Crew® Restroom Floor & Surface SC Non-Acid Disinfectant Cleaner
Cleaning Agents Disinfectants
Non-acid restroom disinfectant cleaner. 
  • No rinsing required for non-food contact surfaces
  • Provides broad spectrum efficacy against common bathroom microoragnisms
  • Kills HIV-1, Staphylococcus aureus, Salmonella choleraesuis, E. coli, MRSA, VRE and other microorganisms
  • Disinfects in up to 400 ppm hard water
  • Deodorizes as it disinfects
  • Fresh scent
Cleans, disinfects and deodorizes in one easy step. Recommended for hard, nonporous, washable surfaces in and out of the restroom.
Utilizes Crew® fresh scent to keep consistent with most Crew® restroom products. Green in color with a fresh scent.