Soft Care® Defend Antibacterial Soap
Supplier: DIVERSEY INC.
100907901
76547-792CT 99.9 USD
76547-792
Soft Care® Defend Antibacterial Soap
Skincare Soaps and Skin Cleansers
Soft Care® Defend(TM/MC) is a highly effective, mild, antibacterial soap. 
  • Safe, quick and effective formulation helps reduce cross-contamination
  • Mild on skin, avoiding excessive cracking and dryness for even the most frequent users
  • NSF E-2 certified for use in food handling establishments
  • Formulated to ensure excellent removal of soil
  • Skin compatibility (Patch) tested
This unique product combines the caring aspect of a gentle-on-skin cosmetic with antiseptic properties.
Formulated with chloroxylenol (PCMX), Soft Care® Defend(TM/MC) is effective at removing soils, rinses quickly and leaves no residue on skin.